Dibutyl malonate ≥99.0%

Supplier: TCI America

M0540-25G M0540-500G
TCM0540-025GEA 148.52 CAD
TCM0540-025G TCM0540-500G
Dibutyl malonate ≥99.0%
Dibutyl malonate

CAS Number: 1190-39-2
MDL Number: MFCD00053795
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 251
Flash Point (°C): 88
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: Malonic Acid Dibutyl Ester

Formula: C₁₁H₂₀O₄
Boiling Pt: 251 °C
Density: 0.98 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00053795
CAS Number: 1190-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf40f1d477c09fc6ece9466c2c923d225249daff&vid=cf40f1d477c09fc6ece9466c2c923d225249daff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR