Variamine blue B sulfate (=4-amino-4'-methoxydiphenylamine sulfate) ≥97.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
N-(4-Methoxyphenyl)-1,4-phenylenediamine sulfate, 4-Amino-4'-methoxydiphenylamine Sulphate, 4-Amino-4'-methoxydiphenylamine sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine sulphate
[for Iron-Titration]
CAS Number: 3169-21-9
MDL Number: MFCD00060258
Molecular Formula: C13H14N2O
Molecular Weight: 312.34
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Other Product Names / Synonyms: 4-Amino-4'-methoxydiphenylamine Sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine Sulfate
Formula:
C₁₃H₁₄N₂O MW: 312.34 g/mol |
MDL Number:
MFCD00060258 CAS Number: 3169-21-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...