Variamine blue B base ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Amino-4'-methoxydiphenylamine, N-(4-Methoxyphenyl)-1,4-phenylenediamine

M0493-1G M0493-25G
TCM0493-001GEA 44.24 CAD
TCM0493-001G TCM0493-025G
Variamine blue B base ≥98.0% (by titrimetric analysis)
Variamine blue B base

[Redox Indicator]
CAS Number: 101-64-4
MDL Number: MFCD00070553
Molecular Formula: C13H14N2O
Molecular Weight: 214.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 101


Other Product Names / Synonyms: 4-Amino-4'-methoxydiphenylamine, N-(4-Methoxyphenyl)-1,4-phenylenediamine

Formula: C₁₃H₁₄N₂O
MW: 214.27 g/mol
Boiling Pt: 210 °C
Melting Pt: 101 °C
MDL Number: MFCD00070553
CAS Number: 101-64-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=686523e8ab99c16fb10b085a529b097922178704&vid=686523e8ab99c16fb10b085a529b097922178704&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR