Myristic anhydride ≥95.0%
Supplier: TCI America
Synonyms:
Myristic anhydride
M0484-250G
M0484-25G
TCM0484-250GEA
1224.82
CAD
TCM0484-250G
TCM0484-025G
Myristic anhydride ≥95.0%
Myristic anhydride
CAS Number: 626-29-9
MDL Number: MFCD00056151
Molecular Formula: C28H54O3
Molecular Weight: 438.74
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 54
Formula:
C₂₈H₅₄O₃ MW: 438.74 g/mol Melting Pt: 54 °C |
MDL Number:
MFCD00056151 CAS Number: 626-29-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...