Sodium myristate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Myristic Acid Sodium Salt
M0483-25G
M0483-500G
M0483-100G
TCM0483-025GEA
96.38
CAD
TCM0483-025G
TCM0483-500G
TCM0483-100G
Sodium myristate ≥98.0% (by GC, titration analysis)
Sodium myristate
CAS Number: 822-12-8
MDL Number: MFCD00042653
Molecular Formula: C14H28O2
Molecular Weight: 250.36
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Product Categories: Materials Chemistry, Anionic Surfactants, Bioscience, Lipids
Other Product Names / Synonyms: Myristic Acid Sodium Salt
Formula:
C₁₄H₂₈O₂ MW: 250.36 g/mol |
MDL Number:
MFCD00042653 CAS Number: 822-12-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...