2-(Methylthio)benzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 2-(Methylthio)benzo[d]thiazole

M0439-25G M0439-500G
TCM0439-025GEA 99.65 CAD
TCM0439-025G TCM0439-500G
2-(Methylthio)benzothiazole ≥98.0%
2-(Methylthio)benzothiazole

CAS Number: 615-22-5
MDL Number: MFCD00005784
Molecular Formula: C8H7NS2
Molecular Weight: 181.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 46
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Mercaptobenzothiazole Methyl Ether, 2-Methylmercaptobenzothiazole

Formula: C₈H₇NS₂
Boiling Pt: 177 °C
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00005784
CAS Number: 615-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c15f6b4e363d59094cf4f9a78b861352af5105c6&vid=c15f6b4e363d59094cf4f9a78b861352af5105c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR