4-Dimethylaminoazobenzene-4'-carboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: 4-Dimethylaminoazobenzene-4-carboxylic Acid, DABCYL acid, Para Methyl Red, p-Methyl Red

M0423-5G M0423-25G
TCM0423-5GEA 200.66 CAD
TCM0423-5G TCM0423-025G
4-Dimethylaminoazobenzene-4'-carboxylic acid ≥97.0%
4-Dimethylaminoazobenzene-4'-carboxylic acid

CAS Number: 6268-49-1
MDL Number: MFCD00058988
Molecular Formula: C15H15N3O2
Molecular Weight: 269.30
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal


Product Categories: Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: p-Methyl Red, Para Methyl Red

Formula: C₁₅H₁₅N₃O₂
MW: 269.30 g/mol
MDL Number: MFCD00058988
CAS Number: 6268-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=263ab09b5e112dd27861e9e601185617c5a41d10&vid=263ab09b5e112dd27861e9e601185617c5a41d10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR