4-Isopropyl-m-cresol ≥99.0% for synthesis
Supplier: TCI America
Synonyms:
4-Isopropyl-3-hydroxytoluene, Biosol, 4-Isopropyl-m-cresol, 4-Isopropyl-3-methylphenol
M0409-25G
M0409-100G
TCM0409-025GEA
62.6
CAD
TCM0409-025G
TCM0409-100G
4-Isopropyl-m-cresol ≥99.0% for synthesis
4-Isopropyl-m-cresol
CAS Number: 3228-02-2
MDL Number: MFCD00010704
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 246
Melting point (°C): 112
Other Product Names / Synonyms: Biosol, 4-Isopropyl-m-cresol
Formula:
C₁₀H₁₄O MW: 150.22 Boiling Pt: 246 °C Melting Pt: 112 °C |
MDL Number:
MFCD00010704 CAS Number: 3228-02-2 EINECS: 221-761-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...