Dimethyl azobenzene-4,4'-dicarboxylate ≥95.0%

Supplier: TCI America

Synonyms: Dimethyl 4,4'-Azodibenzoate, 4,4'-Azodibenzoic Acid Dimethyl Ester, Azobenzene-4,4'-dicarboxylic Acid Dimethyl Ester

A1581-1G A1581-5G
TCA1581-1GEA 263.86 CAD
TCA1581-1G TCA1581-5G
Dimethyl azobenzene-4,4'-dicarboxylate ≥95.0%
Dimethyl azobenzene-4,4'-dicarboxylate

CAS Number: 5320-91-2
MDL Number: MFCD00496622
Molecular Formula: C16H14N2O4
Molecular Weight: 298.30
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 240


Other Product Names / Synonyms: Azobenzene-4,4'-dicarboxylic Acid Dimethyl Ester, 4,4'-Azodibenzoic Acid Dimethyl Ester, Dimethyl 4,4'-Azodibenzoate

Formula: C₁₆H₁₄N₂O₄
MW: 298.30 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00496622
CAS Number: 5320-91-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1b123383df00f889279517bd3801e95a08b6558&vid=e1b123383df00f889279517bd3801e95a08b6558&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR