2-Methyl-4-phenyl-2-butanol ≥99.0%
Supplier: TCI America
Synonyms:
2-Phenylethyldimethylcarbinol, Dimethyl-beta-phenylethylcarbinol
M0398-25G
M0398-500G
M0398-100G
TCM0398-025GEA
80.58
CAD
TCM0398-025G
TCM0398-500G
TCM0398-100G
2-Methyl-4-phenyl-2-butanol ≥99.0%
2-Methyl-4-phenyl-2-butanol
CAS Number: 103-05-9
MDL Number: MFCD00004472
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 30
Other Product Names / Synonyms: Dimethyl-beta-phenylethylcarbinol, 2-Phenylethyldimethylcarbinol
Formula:
C₁₁H₁₆O Flash Pt: 110 °C |
MDL Number:
MFCD00004472 CAS Number: 103-05-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...