3-Carbamyl-1-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Carbamoyl-1-methylpyridinium Chloride, TRIA-662, 3-Carbamoyl-1-methylpyridin-1-ium chloride

M0375-25G M0375-5G
TCM0375-25GEA 998.4 CAD
TCM0375-25G TCM0375-5G
3-Carbamyl-1-methylpyridinium chloride ≥98.0% (by HPLC, titration analysis)
3-Carbamyl-1-methylpyridinium chloride

CAS Number: 1005-24-9
MDL Number: MFCD00060042
Molecular Formula: C7H9ClN2O
Molecular Weight: 172.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 3-Carbamoyl-1-methylpyridinium Chloride

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
MDL Number: MFCD00060042
CAS Number: 1005-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27b4845261feee93209eae49d51fb087b6e31945&vid=27b4845261feee93209eae49d51fb087b6e31945&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR