N-Methylnicotinamide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0374-10G M0374-25G
TCM0374-010GEA 135.56 CAD
TCM0374-010G TCM0374-025G
N-Methylnicotinamide ≥98.0% (by GC, titration analysis)
N-Methylnicotinamide

CAS Number: 114-33-0
MDL Number: MFCD00006384
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 107

Formula: C₇H₈N₂O
Melting Pt: 107 °C
MDL Number: MFCD00006384
CAS Number: 114-33-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa97eeb779872c29a275982313f5392a474b9028&vid=aa97eeb779872c29a275982313f5392a474b9028&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR