Leuco crystal violet 98,0

Supplier: TCI America

Synonyms: 4,4',4''-Methanetriyltris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane, 4,4',4''-Methylidynetris(N,N-dimethylaniline)

M0344-250G M0344-25G
TCM0344-250GEA 518.24 CAD
TCM0344-250G TCM0344-025G
Leuco crystal violet 98,0
Leuco crystal violet

CAS Number: 603-48-5
MDL Number: MFCD00008314
Molecular Formula: C25H31N3
Molecular Weight: 373.54
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 178


Other Product Names / Synonyms: 4,4',4''-Methylidynetris(N,N-dimethylaniline), Tris(4-dimethylaminophenyl)methane

Formula: C₂₅H₃₁N₃
MW: 373.54 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00008314
CAS Number: 603-48-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36a81690da2eee12a6f8c41144011b74ab982cff&vid=36a81690da2eee12a6f8c41144011b74ab982cff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR