1-Methylguanidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
M0336-25G
M0336-5G
TCM0336-025GEA
271.76
CAD
TCM0336-025G
76220-226
1-Methylguanidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
1-Methylguanidine hydrochloride
CAS Number: 21770-81-0
MDL Number: MFCD00012576
Molecular Formula: C2H7N3
Molecular Weight: 109.56
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Formula:
C₂H₈ClN₃ |
MDL Number:
MFCD00012576 CAS Number: 21770-81-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...