1-Acetyladamantane ≥98.0%

Supplier: TCI America

A1571-25G A1571-5G
TCA1571-25GEA 349.18 CAD
TCA1571-25G TCA1571-5G
1-Acetyladamantane ≥98.0%
1-Acetyladamantane

CAS Number: 1660-04-4
MDL Number: MFCD00074739
Molecular Formula: C12H18O
Molecular Weight: 178.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 108


Other Product Names / Synonyms: 1-Adamantyl Methyl Ketone, Methyl 1-Adamantyl Ketone

Formula: C₁₂H₁₈O
Melting Pt: 54 °C
Flash Pt: 108 °C
MDL Number: MFCD00074739
CAS Number: 1660-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e1f307e50e85e3d85a4bc9c249ea683bd033287&vid=3e1f307e50e85e3d85a4bc9c249ea683bd033287&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR