Tridecyl methacrylate (mixture of branched chain isomers) stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid Tridecyl Ester

M0264-25ML M0264-500ML
TCM0264-025MLEA 47.4 CAD
TCM0264-025ML TCM0264-500ML
Tridecyl methacrylate (mixture of branched chain isomers) stabilised
Tridecyl methacrylate (mixture of branched chain isomers)

(stabilized with MEHQ)
CAS Number: 2495-25-2
MDL Number: MFCD00059220
Molecular Formula: C17H32O2
Molecular Weight: 268.44
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 150
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Methacrylic Acid Tridecyl Ester

Formula: C₁₇H₃₂O₂
MW: 268.44 g/mol
Density: 0.88 (20 °C)
Flash Pt: 150 °C
MDL Number: MFCD00059220
CAS Number: 2495-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3c9ba27be5490b096ab627226858288ea6a6956&vid=b3c9ba27be5490b096ab627226858288ea6a6956&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR