2,2'-Methylenebis(6-tert-butyl-p-cresol) ≥99.0%
Supplier: TCI America
Synonyms:
2,2'-Methylenebis(4-methyl-6-tert-butylphenol), 2,2-Methylenebis(6-tert-butyl-p-cresol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 2,2'-Methylenebis(6-tert-butyl-4-methylphenol), 3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5'-dimethyldiphenylmethane
CAS Number: 119-47-1
MDL Number: MFCD00043641
Molecular Formula: C23H32O2
Molecular Weight: 340.51
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 130
Flash Point (°C): 195
Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5'-dimethyldiphenylmethane, 2,2'-Methylenebis(6-tert-butyl-4-methylphenol)
Formula:
C₂₃H₃₂O₂ MW: 340.51 g/mol Melting Pt: 130 °C Flash Pt: 195 °C |
MDL Number:
MFCD00043641 CAS Number: 119-47-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...