2,2'-Methylenebis(6-tert-butyl-4-ethylphenol) ≥98.0%

Supplier: TCI America

M0215-500G M0215-25G
TCM0215-500GEA 221.52 CAD
TCM0215-500G TCM0215-025G
2,2'-Methylenebis(6-tert-butyl-4-ethylphenol) ≥98.0%
2,2'-Methylenebis(6-tert-butyl-4-ethylphenol)

CAS Number: 88-24-4
MDL Number: MFCD00026296
Molecular Formula: C25H36O2
Molecular Weight: 368.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125


Product Categories: Materials Chemistry, Resin Additives, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methane, 3,3'-Di-tert-butyl-5,5'-diethyl-2,2'-dihydroxydiphenylmethane, 2,2'-Methylenebis(4-ethyl-6-tert-butylphenol)

Formula: C₂₅H₃₆O₂
Melting Pt: 125 °C
MDL Number: MFCD00026296
CAS Number: 88-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44bc9bf5f02f4aec42f2985d74087c946d20d899&vid=44bc9bf5f02f4aec42f2985d74087c946d20d899&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR