3-Methylcyclohexanol ≥98.0%

Supplier: TCI America

M0195-25ML M0195-500ML
TCM0195-025MLEA 57.84 CAD
TCM0195-025ML TCM0195-500ML
3-Methylcyclohexanol ≥98.0%
3-Methylcyclohexanol

CAS Number: 591-23-1
MDL Number: MFCD00001446
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 62
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 3-Methylhexalin

Formula: C₇H₁₄O
Boiling Pt: 171…173 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.92 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00001446
CAS Number: 591-23-1
EINECS: 209-709-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9fdf6db7f8e4cab7731c5bdadf428055c538478&vid=9c53cf3192a4f7e3b8a412bf8239772fc89acc15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR