6-Methoxy-m-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-Amino-4-methoxytoluene, 2-Amino-4-methylanisole, 1-Amino-2-methoxy-5-methylbenzene, 5-Methyl-o-anisidine, p-Cresidine, 2-Methoxy-5-methylaniline

M0115-25G M0115-500G
TCM0115-025GEA 41.08 CAD
TCM0115-025G TCM0115-500G
6-Methoxy-m-toluidine ≥98.0% (by GC, titration analysis)
6-Methoxy-m-toluidine

CAS Number: 120-71-8
MDL Number: MFCD00007815
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 52
Flash Point (°C): 111


Other Product Names / Synonyms: p-Cresidine, 6-Methoxy-m-toluidine, 5-Methyl-o-anisidine

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 235 °C
Melting Pt: 52 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 111 °C
MDL Number: MFCD00007815
CAS Number: 120-71-8
EINECS: 204-419-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48205ce1615e67a46a0378902fd8ede9e9da7ff9&vid=48205ce1615e67a46a0378902fd8ede9e9da7ff9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR