Methyl purple in solution

Supplier: TCI America

Synonyms: Fleisher's Indicator Solution

M0112-25ML M0112-500ML
TCM0112-025MLEA 68.57 CAD
TCM0112-025ML TCM0112-500ML
Methyl purple in solution
Methyl purple

pH 4.8 (Purple) - pH 5.4 ( Gray) - pH 5.8 (Green)
MDL Number: MFCD00070843
Form: Liquid
Color: Deep Green
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Fleisher's Indicator Solution

Formula: C67H62N8Na4O14S3
MDL Number: MFCD00070843
CAS Number: 1340-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR