Isobutyl methacrylate polymer
Supplier: TCI America
M0086-500G
M0086-25G
TCM0086-500GEA
186.44
CAD
TCM0086-500G
TCM0086-025G
Isobutyl methacrylate polymer
Isobutyl methacrylate polymer
CAS Number: 9011-15-8
MDL Number: MFCD00008931
Molecular Formula: C8H14O2
Form: Crystal
Color: White
Other Product Names / Synonyms: Methacrylic Acid Isobutyl Ester Polymer
Formula:
C₈H₁₄O₂ |
MDL Number:
MFCD00008931 CAS Number: 9011-15-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Viscosity | 100.0 to 160.0 mPa-s (40 wt%, Toluene, 25 deg-C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...