m-Cresol purple for synthesis

Supplier: TCI America

Synonyms: m-Cresolsulfonphthalein, 2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 2',2''-Dimethylphenolsulfonphthalein

M0074-1G M0074-25G
TCM0074-001GEA 80.77 CAD
TCM0074-001G TCM0074-025G
m-Cresol purple for synthesis
m-Cresol purple

CAS Number: 2303-01-7
MDL Number: MFCD00005871
Molecular Formula: C21H18O5S
Molecular Weight: 382.43
Form: Crystal
Color: Yellow Green


Other Product Names / Synonyms: m-Cresol Purple

Formula: C₂₁H₁₈O₅S
MW: 382.43 g/mol
MDL Number: MFCD00005871
CAS Number: 2303-01-7
EINECS: 218-960-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128c7f9762c89b34e8de4b1246a2030d7225e05d&vid=128c7f9762c89b34e8de4b1246a2030d7225e05d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR