Mesotartaric acid monohydrate ≥90.0% (by HPLC)

Supplier: TCI America

Synonyms: meso-Tartaric Acid

M0070-5G M0070-25G
TCM0070-005GEA 259.12 CAD
TCM0070-005G TCM0070-025G
Mesotartaric acid monohydrate ≥90.0% (by HPLC)
Mesotartaric acid monohydrate

CAS Number: 5990-63-6
MDL Number: MFCD00063641
Molecular Formula: C4H6O6
Molecular Weight: 150.09
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Melting point (°C): 75


Other Product Names / Synonyms: meso-Tartaric Acid

Formula: C₄H₆O₆
MW: 150.09 g/mol
Melting Pt: 75 °C
MDL Number: MFCD00063641
CAS Number: 5990-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deef5f48976b50948d8697cb8614c7c931f9d240&vid=deef5f48976b50948d8697cb8614c7c931f9d240&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR