D-(+)-Melibiose monohydrate ≥99.0% (by HPLC)
Supplier: TCI America
M0050-10G
M0050-1G
TCM0050-010GEA
183.28
CAD
TCM0050-010G
TCM0050-001G
D-(+)-Melibiose monohydrate ≥99.0% (by HPLC)
D-(+)-Melibiose monohydrate
CAS Number: 585-99-9
MDL Number: MFCD00198188
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 182
Specific rotation [a]20/D: 136 deg (C=4, H2O)
Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Disaccharides
Other Product Names / Synonyms: 6-O-alpha-D-Galactopyranosyl-D-glucopyranose
Formula:
C₁₂H₂₂O₁₁·H₂O Melting Pt: 182 °C |
MDL Number:
MFCD00198188 CAS Number: 66009-10-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...