D-(+)-Melezitose hydrate ≥99.0%
Supplier: TCI America
M0049-25G
M0049-5G
TCM0049-025GEA
489.33
CAD
TCM0049-025G
TCM0049-005G
D-(+)-Melezitose hydrate ≥99.0%
D-(+)-Melezitose hydrate
CAS Number: 207511-10-2
MDL Number: MFCD00149448
Molecular Formula: C18H32O16
Molecular Weight: 504.44
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: 88.5 deg (C=4, H2O)
Formula:
C₁₈H₃₄O₁₇ Melting Pt: 153 °C |
MDL Number:
MFCD00149448 CAS Number: 207511-10-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...