(±)-Ethyl mandelate ≥95.0%

Supplier: TCI America

M0039-25G M0039-500G
TCM0039-025GEA 56.25 CAD
TCM0039-025G TCM0039-500G
(±)-Ethyl mandelate ≥95.0%
(±)-Ethyl mandelate

CAS Number: 774-40-3
MDL Number: MFCD00004494
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 133
Flash Point (°C): 110
Freezing point (°C): 26


Other Product Names / Synonyms: DL-Mandelic Acid Ethyl Ester

Formula: C₁₀H₁₂O₃
Boiling Pt: 133 °C
Flash Pt: 110 °C
MDL Number: MFCD00004494
CAS Number: 774-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ced04a98b8e186fbf56ae6323ef36b0e1a16c080&vid=ced04a98b8e186fbf56ae6323ef36b0e1a16c080&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR