(±)-Mandelic acid ≥99.0%
Supplier: TCI America
Synonyms:
DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
M0038-25G
M0038-500G
TCM0038-025GEA
48.98
CAD
TCM0038-025G
TCM0038-500G
(±)-Mandelic acid ≥99.0%
(±)-Mandelic acid
CAS Number: 90-64-2
MDL Number: MFCD00064250
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 121
Other Product Names / Synonyms: DL-Amygdalic Acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 322 Melting Pt: 121 °C |
MDL Number:
MFCD00064250 CAS Number: 90-64-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...