DL-Malic acid disodium salt ≥98.0%
Supplier: TCI America
Synonyms:
Di-Sodium Dl-Malate, DL-Malic acid disodium salt, DL-Apple acid disodium salt, Disodium DL-Malate hydrate
M0026-500G
M0026-25G
TCM0026-500GEA
127.98
CAD
TCM0026-500G
TCM0026-025G
DL-Malic acid disodium salt ≥98.0%
DL-Malic acid disodium salt
CAS Number: 676-46-0
MDL Number: MFCD00012466
Molecular Formula: C4H6O5
Molecular Weight: 178.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: DL-Apple Acid Disodium Salt, DL-Malic Acid Disodium Salt
Formula:
C₄H₄Na₂O₅ |
MDL Number:
MFCD00012466 CAS Number: 676-46-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...