Diethyl-DL-malate ≥98.0%

Supplier: TCI America

M0024-25G M0024-500G
TCM0024-025GEA 48.98 CAD
TCM0024-025G TCM0024-500G
Diethyl-DL-malate ≥98.0%
Diethyl-DL-malate

CAS Number: 7554-12-3
MDL Number: MFCD00048556
Molecular Formula: C8H14O5
Molecular Weight: 190.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 85
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: DL-Apple Acid Diethyl Ester, DL-Malic Acid Diethyl Ester

Formula: C₈H₁₄O₅
Boiling Pt: 255 °C
Density: 1.13 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00048556
CAS Number: 7554-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba489f015daf1abba82bb500d3e4e7a9131336d4&vid=ba489f015daf1abba82bb500d3e4e7a9131336d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR