Dibutyl-DL-malate ≥95.0%

Supplier: TCI America

Synonyms: DL-Apple Acid Dibutyl Ester, DL-Malic Acid Dibutyl Ester

M0023-500G M0023-25G
TCM0023-500GEA 1230.82 CAD
TCM0023-500G TCM0023-025G
Dibutyl-DL-malate ≥95.0%
Dibutyl-DL-malate

CAS Number: 6280-99-5
MDL Number: MFCD00059251
Molecular Formula: C12H22O5
Molecular Weight: 246.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: DL-Apple Acid Dibutyl Ester, DL-Malic Acid Dibutyl Ester

Formula: C₁₂H₂₂O₅
Density: 1.04 (20 °C)
MDL Number: MFCD00059251
CAS Number: 6280-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e891a93c5bbb133161b7cfc6aef114201cd071ba&vid=e891a93c5bbb133161b7cfc6aef114201cd071ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR