Disodium maleate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Maleic acid disodium salt

M0014-25G M0014-500G
TCM0014-025GEA 59.87 CAD
TCM0014-025G TCM0014-500G
Disodium maleate ≥99.0% (by titrimetric analysis)
Disodium maleate

CAS Number: 371-47-1
MDL Number: MFCD00013059
Molecular Formula: C4H4O4
Molecular Weight: 160.04
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Other Product Names / Synonyms: Maleic Acid Disodium Salt

Formula: C₄H₂Na₂O₄
MDL Number: MFCD00013059
CAS Number: 371-47-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0a2f51c460e951b534173774a68665c1b11bb6a&vid=e0a2f51c460e951b534173774a68665c1b11bb6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR