Bis(2-ethylhexyl)maleate ≥93.0%

Supplier: TCI America

M0011-25ML M0011-500ML
TCM0011-025MLEA 48.98 CAD
TCM0011-025ML TCM0011-500ML
Bis(2-ethylhexyl)maleate ≥93.0%
Bis(2-ethylhexyl)maleate

CAS Number: 142-16-5
MDL Number: MFCD00027250
Molecular Formula: C20H36O4
Molecular Weight: 340.50
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Melting point (°C): -60
Flash Point (°C): 180
Specific Gravity (20/20): 0.94


Product Categories: Materials Chemistry, Resin Additives, Copolymerization Monomers
Other Product Names / Synonyms: Di(2-ethylhexyl) Maleate, Dioctyl Maleate, Maleic Acid Di(2-ethylhexyl) Ester, Maleic Acid Dioctyl Ester

Formula: C₂₀H₃₆O₄
Boiling Pt: 173 °C
Melting Pt: –60 °C
Density: 0.94 (20 °C)
Flash Pt: 180 °C
MDL Number: MFCD00027250
CAS Number: 142-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3657abc9389b146f104c582d273c6f129f5478c7&vid=3657abc9389b146f104c582d273c6f129f5478c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR