1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane) ≥98.0% for synthesis

Supplier: TCI America

Synonyms: 1-Aza-12-crown 4-Ether, 1,4,7-Trioxa-10-azacyclododecane

A1552-1G A1552-250MG
TCA1552-1GEA 489.33 CAD
TCA1552-1G TCA1552-250MG
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane) ≥98.0% for synthesis
1-Aza-12-crown-4 (1,4,7-Trioxa-10-azacyclododecane)

CAS Number: 41775-76-2
MDL Number: MFCD00134269
Molecular Formula: C8H17NO3
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 58


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7-Trioxa-10-azacyclododecane

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Melting Pt: 58 °C
MDL Number: MFCD00134269
CAS Number: 41775-76-2
EINECS: 255-549-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c844701cdbc464d8e7b7007b6c39ec03bfca4483&vid=c844701cdbc464d8e7b7007b6c39ec03bfca4483&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR