Magnesium acetylacetonate ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Magnesium bis(2,4-pentanedionato-O,O'), Bis(2,4-pentanedionato-O,O')magnesium, Magnesiumacetylacetonate, Magnesium 2,4-pentanedionate

M0001-25G M0001-5G
TCM0001-025GEA 267.02 CAD
TCM0001-025G TCM0001-5G
Magnesium acetylacetonate ≥98.0% for synthesis
Magnesium acetylacetonate

CAS Number: 14024-56-7
MDL Number: MFCD00013497
Molecular Formula: C10H14MgO4
Molecular Weight: 222.52
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 276


Other Product Names / Synonyms: Acetylacetone Magnesium(II) Salt, Magnesium(II) Acetylacetonate

Formula: C₁₀H₁₄MgO₄
Melting Pt: 276 °C
MDL Number: MFCD00013497
CAS Number: 14024-56-7
EINECS: 237-857-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f53c4b986649458f395281f8a12a3603c7559e05&vid=f53c4b986649458f395281f8a12a3603c7559e05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR