D-Leucine methyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-D-Leu-OMe.HCl

L0198-1G L0198-5G
TCL0198-1GEA 52.14 CAD
TCL0198-1G TCL0198-5G
D-Leucine methyl ester hydrochloride ≥98.0%
D-Leucine methyl ester hydrochloride

CAS Number: 5845-53-4
MDL Number: MFCD00066115
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 151
Specific rotation [a]20/D: -13 deg (C=2, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Leu-OMe.HCl

Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 151 °C
MDL Number: MFCD00066115
CAS Number: 5845-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b4e15d8e035eddc52f659c65bdeceb2a4864de1&vid=8b4e15d8e035eddc52f659c65bdeceb2a4864de1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR