H-D-Leu-OtBu HCl ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: H-D-Leu-OtBu·HCl, (R)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride

L0196-1G L0196-5G
TCL0196-1GEA 56.88 CAD
TCL0196-1G TCL0196-5G
H-D-Leu-OtBu HCl ≥98.0% (by total nitrogen and titration analysis)
H-D-Leu-OtBu HCl

CAS Number: 13081-32-8
MDL Number: MFCD00083642
Molecular Formula: C10H21NO2
Molecular Weight: 223.74
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Specific rotation [a]20/D: -19 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Leu-OtBu.HCl

Formula: C₁₀H₂₂ClNO₂
MW: 223.7402 g/mol
MDL Number: MFCD00083642
CAS Number: 13081-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1f6c21135244cacc4980247704e00c26f783e28&vid=a1f6c21135244cacc4980247704e00c26f783e28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR