L-Leucine benzyl ester-p-toluenesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
L-Leucine benzyl ester p-toluenesulfonate salt, H-Leu-OBzl.Tos-OH, L-Leucine benzyl ester p-toluenesulphonate salt, H-Leu-OBzl·TosOH
CAS Number: 1738-77-8
MDL Number: MFCD00066117
Molecular Formula: C13H19NO2
Molecular Weight: 393.50
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158
Specific rotation [a]20/D: 5 deg (C=2, DMF)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Leu-OBzl.Tos-OH
Formula:
C₁₃H₁₉NO₂ MW: 393.50 g/mol Melting Pt: 158 °C |
MDL Number:
MFCD00066117 CAS Number: 1738-77-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...