L-(+)-Methyl-tert-leucinate hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl
CAS Number: 63038-27-7
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Specific rotation [a]20/D: 16.5 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester Hydrochloride, Methyl (S)-2-Amino-3,3-dimethylbutyrate Hydrochloride, H-Tle-OMe.HCl
Formula:
C₇H₁₅NO₂·HCl |
MDL Number:
MFCD00040604 CAS Number: 63038-27-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...