L-tert-leucine methylamide ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: L-tert-Leucylaminomethane, (S)-2-Amino-N,3,3-trimethylbutanamide

L0168-5G L0168-1G
TCL0168-5GEA 731.54 CAD
TCL0168-5G TCL0168-1G
L-tert-leucine methylamide ≥96.0% (by titrimetric analysis)
L-tert-leucine methylamide

CAS Number: 89226-12-0
MDL Number: MFCD00673329
Molecular Formula: C7H16N2O
Molecular Weight: 144.22
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Specific rotation [a]20/D: 55 deg (C=0.5, H2O)


Other Product Names / Synonyms: L-tert-Leucylaminomethane

Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
MDL Number: MFCD00673329
CAS Number: 89226-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93f3c8e16bab0c392358bd17be2fb104089c1d8d&vid=93f3c8e16bab0c392358bd17be2fb104089c1d8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR