L-tert-Leucinol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

L0160-5G L0160-1G
TCL0160-005GEA 295.46 CAD
TCL0160-005G TCL0160-001G
L-tert-Leucinol ≥97.0% (by GC, titration analysis)
L-tert-Leucinol

CAS Number: 112245-13-3
MDL Number: MFCD00192250
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 130
Flash Point (°C): 90
Freezing point (°C): 33
Specific rotation [a]20/D: 38 deg (C=1.5, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-Amino-3,3-dimethylbutanol

Formula: C₆H₁₅NO
Boiling Pt: 130 °C
Flash Pt: 90 °C
MDL Number: MFCD00192250
CAS Number: 112245-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db1385e32dca27d132f1ca1b91c5d944a887046c&vid=db1385e32dca27d132f1ca1b91c5d944a887046c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR