2-Ethylphenyl acetate ≥97.0%

Supplier: TCI America

A1549-5G A1549-25G
TCA1549-005GEA 86.77 CAD
TCA1549-005G TCA1549-025G
2-Ethylphenyl acetate ≥97.0%
2-Ethylphenyl acetate

CAS Number: 3056-59-5
MDL Number: MFCD02093429
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Acetic Acid 2-Ethylphenyl Ester

Formula: C₁₀H₁₂O₂
Density: 1.03 (20 °C)
MDL Number: MFCD02093429
CAS Number: 3056-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08f311899ada1f3eecc0753a0efd68c5397d9a3f&vid=08f311899ada1f3eecc0753a0efd68c5397d9a3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR