γ-Linolenic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

L0152-100MG L0152-1G
TCL0152-100MGEA 160.64 CAD
TCL0152-100MG TCL0152-1G
γ-Linolenic acid ≥98.0% (by GC, titration analysis)
γ-Linolenic acid

CAS Number: 506-26-3
MDL Number: MFCD00065718
Molecular Formula: C18H30O2
Molecular Weight: 278.44
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: cis,cis,cis-6,9,12-Octadecatrienoic Acid

Formula: C₁₈H₃₀O₂
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00065718
CAS Number: 506-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40c96df694ba55be5628c9557cb873e96ce9cfea&vid=40c96df694ba55be5628c9557cb873e96ce9cfea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR