Linoleic anhydride ≥90.0%

Supplier: TCI America

Synonyms: cis,cis-9,12-Octadecadienoic Anhydride

L0145-1G L0145-5G
TCL0145-001GEA 86.9 CAD
TCL0145-001G TCL0145-5G
Linoleic anhydride ≥90.0%
Linoleic anhydride

CAS Number: 24909-68-0
MDL Number: MFCD00010456
Molecular Formula: C36H62O3
Molecular Weight: 542.89
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C


Other Product Names / Synonyms: cis,cis-9,12-Octadecadienoic Anhydride

Formula: C₃₆H₆₂O₃
MW: 542.89 g/mol
Density: 0.90 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00010456
CAS Number: 24909-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afb54c0b3d4190ca1b5d291743145b3550f628f6&vid=afb54c0b3d4190ca1b5d291743145b3550f628f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR