L-Lysine ≥97.0% for synthesis

Supplier: TCI America

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

L0129-5G L0129-25G
TCL0129-005GEA 59.25 CAD
TCL0129-005G TCL0129-025G
L-Lysine ≥97.0% for synthesis
L-Lysine

CAS Number: 56-87-1
MDL Number: MFCD00064433
Molecular Formula: C6H14N2O2
Molecular Weight: 146.19
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 212
Specific rotation [a]20/D: 26 deg (C=2, 5mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Other Plant Hormones, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Lys-OH

Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Boiling Pt: 290
Melting Pt: 212 °C
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df1015e857d6385199510555ddf26bb56cc13dee&vid=df1015e857d6385199510555ddf26bb56cc13dee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR