DL-Leucic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

L0104-500G L0104-25G
TCL0104-500GEA 1499.42 CAD
TCL0104-500G TCL0104-025G
DL-Leucic acid ≥98.0% (by GC, titration analysis)
DL-Leucic acid

CAS Number: 498-36-2
MDL Number: MFCD00004246
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 75


Other Product Names / Synonyms: 2-Hydroxy-4-methylvaleric Acid

Formula: C₆H₁₂O₃
Melting Pt: 75 °C
MDL Number: MFCD00004246
CAS Number: 498-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6917fa2fbf256ef86ce1d70f288a446f2a03d56f&vid=6917fa2fbf256ef86ce1d70f288a446f2a03d56f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR