Sodium ligninsulfonate
Supplier: TCI America
L0098-25G
L0098-500G
TCL0098-025GEA
58.46
CAD
TCL0098-025G
TCL0098-500G
Sodium ligninsulfonate
Sodium ligninsulfonate
CAS Number: 8061-51-6
MDL Number: MFCD00135572
Form: Crystal
Color: Deep Reddish Yellow
Other Product Names / Synonyms: Ligninsulfonic Acid Sodium Salt
MDL Number:
MFCD00135572 CAS Number: 8061-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...