Copper(I) acetate ≥93.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A1540-25G A1540-5G
TCA1540-25GEA 571.96 CAD
TCA1540-25G TCA1540-005G
Copper(I) acetate ≥93.0% (by titrimetric analysis)
Copper (I) acetate

CAS Number: 598-54-9
MDL Number: MFCD00058908
Molecular Formula: C2H4O2
Molecular Weight: 122.59
Purity/Analysis Method: >93.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Metal Catalysts & Ligands (Click Chemistry)
Other Product Names / Synonyms: Acetic Acid Copper(I) Salt

Formula: C₂H₃CuO₂
Melting Pt: 250 °C
MDL Number: MFCD00058908
CAS Number: 598-54-9
EINECS: 209-938-7
Merck Index: 12,02728
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d58cd7e61ee13787425a23ea52b5bdb8bdb7af1&vid=7d58cd7e61ee13787425a23ea52b5bdb8bdb7af1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR