Vinyl laurate ≥99.0% stabilized

Supplier: TCI America

L0090-25ML L0090-500ML
TCL0090-025MLEA 33.18 CAD
TCL0090-025ML TCL0090-500ML
Vinyl laurate ≥99.0% stabilized
Vinyl laurate

(stabilized with MEHQ)
CAS Number: 2146-71-6
MDL Number: MFCD00048421
Molecular Formula: C14H26O2
Molecular Weight: 226.36
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 285
Flash Point (°C): 136
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Lauric Acid Vinyl Ester

Formula: C₁₄H₂₆O₂
Boiling Pt: 285 °C
Density: 0.87 (20 °C)
Flash Pt: 136 °C
MDL Number: MFCD00048421
CAS Number: 2146-71-6
EINECS: 218-414-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e39cdef48c1aea6f60cfd54a5f4b51fc438e3a&vid=02e39cdef48c1aea6f60cfd54a5f4b51fc438e3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR