Lauramide ≥96.0%
Supplier: TCI America
L0077-25G
L0077-500G
L0077-100G
TCL0077-025GEA
137.71
CAD
TCL0077-025G
TCL0077-500G
TCL0077-100G
Lauramide ≥96.0%
Lauramide
CAS Number: 1120-16-7
MDL Number: MFCD00025532
Molecular Formula: C12H25NO
Molecular Weight: 199.34
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 99
Other Product Names / Synonyms: Dodecanamide
Formula:
C₁₂H₂₅NO Boiling Pt: 200 °C Melting Pt: 99 °C |
MDL Number:
MFCD00025532 CAS Number: 1120-16-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...